Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl 6-Aminohexanoate p-Toluenesulfonate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
1926-86-9
C19H33NO5S
387.535
MFCD00059852
RVALTWBPTNLNAV-UHFFFAOYSA-N
6-Aminohexanoic Acid Hexyl Ester p-Toluenesulfonate
44630052
hexyl 6-aminohexanoate;4-methylbenzenesulfonic acid
CCCCCCOC(=O)CCCCCN.CC1=CC=C(C=C1)S(=O)(=O)O
1926-86-9
C19H33NO5S
387.535
MFCD00059852
RVALTWBPTNLNAV-UHFFFAOYSA-N
6-Aminohexanoic Acid Hexyl Ester p-Toluenesulfonate
44630052
hexyl 6-aminohexanoate;4-methylbenzenesulfonic acid
CCCCCCOC(=O)CCCCCN.CC1=CC=C(C=C1)S(=O)(=O)O
Melting Point 96°C
Quantity 5g
Color White
Formula Weight 387.54
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T,N)
Chemical Name or Material Hexyl 6-Aminohexanoate p-Toluenesulfonate