Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
1L
Packaging:
Glass bottle
111-26-2
C6H15N
101.19
MFCD00008240
BMVXCPBXGZKUPN-UHFFFAOYSA-N
hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
8102
CHEBI:5712
hexan-1-amine
CCCCCCN
111-26-2
C6H15N
101.19
MFCD00008240
BMVXCPBXGZKUPN-UHFFFAOYSA-N
hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
8102
CHEBI:5712
hexan-1-amine
CCCCCCN
Viscosity 0.83 mPa.s (20°C)
Water 0.5% max. (K.F.)
Specific Gravity 0.75
Infrared Spectrum Authentic
Boiling Point 130.0°C
Packaging Glass bottle
Flash Point 27°C
Melting Point -19.0°C
Refractive Index 1.4170 to 1.4190
Quantity 1L
Color Colorless to Yellow
Density 0.7500g/mL
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)5NH2
Beilstein 04, 188
Fieser 07,170
Solubility Information Solubility in water: 14g/L (20°C).
Formula Weight 101.19
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Hexylamine, 99%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 203-851-8

RTECSNumber : MQ4540000

TSCA : TSCA