HexylHIBO, Tocris

Group I mGlu antagonist

Manufacturer: R&D Systems 1749/50

Quantity 50mg

 View more versions of this product

Catalog No. 17-495-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C12H20N2O4
    Formula Weight 256.3
    Synonym α-Amino-4-hexyl-2, 3-dihydro-3-oxo-5-isoxazolepropanoic acid
    Chemical Name or Material HexylHIBO

    Specifications

    Formula Weight 256.3
    Quantity 50mg
    Assay Percent Range >98%
    Molecular Formula C12H20N2O4
    CAS 334887-43-3
    Synonym α-Amino-4-hexyl-2, 3-dihydro-3-oxo-5-isoxazolepropanoic acid
    Recommended Storage Store at Room Temperature
    Chemical Name or Material HexylHIBO

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit