Learn More
HexylHIBO, Tocris
Group I mGlu antagonist
Manufacturer:  R&D Systems 1749/10
Quantity 10mg
 View more versions of this product
Catalog No. 17-491-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
256.3
HexylHIBO
C12H20N2O4
α-Amino-4-hexyl-2, 3-dihydro-3-oxo-5-isoxazolepropanoic acid
Specifications
10mg
>98%
256.3
HexylHIBO
334887-43-3
C12H20N2O4
α-Amino-4-hexyl-2, 3-dihydro-3-oxo-5-isoxazolepropanoic acid
Safety and Handling
Recommended Storage : Store at Room Temperature