Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEY1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523484
Description
HEY1 Polyclonal Antibody for Western Blot
Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.
Specifications
HEY1 | |
Polyclonal | |
Unconjugated | |
HEY1 | |
AI316788; AI414254; basic helix-loop-helix protein OAF1; basic helix-loop-helix transcription factor; BC8; bHLH protein Hesr-1/Hey1; bHLHb31; cardiovascular basic helix-loop-helix factor 2; cardiovascular helix-loop-helix factor 2; CHF2; CHF-2; class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif protein 1; hairy-related transcription factor 1; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 1; Hesr1; HESR-1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; HEY1; hey1 protein; hHRT1; HRT1; HRT-1; id:ibd1292; MGC1274; mHRT1; OAF1; zgc:110572 | |
Rabbit | |
Protein A | |
RUO | |
15213 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q9WV93 | |
HEY1 | |
A portion of amino acids 50-100 of mouse Hey1. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction