Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEY2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599640
Description
Antibody detects endogenous levels of total HRT2.
Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
Specifications
HEY2 | |
Polyclonal | |
Unconjugated | |
Hey2 | |
basic helix-loop-helix transcription factor; bHLHb32; cardiovascular basic helix-loop-helix factor 1; cardiovascular helix-loop-helix factor 1; Chf1; Class B basic helix-loop-helix protein 32; gridlock; GRL; h hHRT2; hairy and enhancer of split related 2; hairy and enhancer of split-related protein 2; hairy/enhancer-of-split related with YRPW motif 2; hairy/enhancer-of-split related with YRPW motif protein 2; Hairy-E(spl)-related with YRPW motif 2; hairy-related transcription factor 2; hCHF1; HERP; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 2; hesr2; HESR-2; hes-related family bHLH transcription factor with YRPW motif 2; HES-related repressor protein 1; HES-related repressor protein 2; hey2; hHRT2; Hrt2; HRT-2; mHRT2; protein gridlock; Protein gridlock homolog; zgc:136746 | |
Rabbit | |
Affinity chromatography | |
RUO | |
15214, 155430, 23493 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9QUS4, Q9UBP5 | |
Hey2 | |
A synthesized peptide derived from human HEY2(Accession Q9UBP5), corresponding to amino acid residues S27-T77. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction