Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | HEY2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HEY2 Polyclonal specifically detects HEY2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HEY2 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BHLHB32, bHLHb32HES-related repressor protein 2, cardiovascular basic helix-loop-helix factor 1, Cardiovascular helix-loop-helix factor 1, CHF1, Class B basic helix-loop-helix protein 32, GRLGRIDLOCK, Hairy and enhancer of split-related protein 2, hairy/enhancer-of-split related with YRPW motif 2, hairy/enhancer-of-split related with YRPW motif protein 2, Hairy-related transcription factor 2, hCHF1, HERP, HERP1hHRT2, HESR2, HESR-2, HES-related repressor protein 1, HRT2, HRT-2, MGC10720, Protein gridlock homolog | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HEY2 (NP_036391.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
23493 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title