Learn More
HF scientific Micro 100 Laboratory Turbidimeter
Ideal for municipal water and wastewater facilities, environmental, commercial and industrial laboratories
Manufacturer:  HF scientific 19950
 View more versions of this product
Catalog No. 15-393-81
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
Micro 100 Laboratory Turbidimeter
±0.01 NTU
10 x 10.7 x 4.75 in. (25.4 x 23.7 x 12.1cm)
Complete set of calibration standards (0.02, 10 and 1000 NTU), three measuring cuvettes with light shield cap, operator's manual and a universal power adapter that operates at any voltage between 100/240VAC
0 to 1000 NTU
12.1cm
25.4cm
≤6 sec.
32° to 122°F
100/240V
1.32kg
23.7cm
CE, US EPA
0.01, 0.1, 1 NTU
ProCal calibration standards
Ideal for municipal water and wastewater facilities, environmental, commercial and industrial laboratories
Nephelometric non-ratio
4.75 in.
10 in.
Quick connect Tungsten filament or infrared
27 to 30mL
0° to 50°C
2.9 lb.
10.7 in.
1 Year