HF scientific Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
Manufacturer: HF scientific 10502
 View more versions of this product
Catalog No. 15-393-151
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For testing total chlorine
    For Use With (Equipment) HF scientific and Hach™ photometers
    No. of Tests 100.0
    Sample Size 5mL
    Product Type Dispenser
  • Description & Specifications
Related Products
Product Content Feedback