Learn More
HGF Mouse anti-Human, Bovine, Alexa Fluor™ 350, Clone: MM0326-5D20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212355AF350
Catalog No. NP2123A30
Add to cart
Description
Description
HGF Monoclonal antibody specifically detects HGF in Human,Bovine samples. It is validated for Western Blot.
Specifications
Specifications
HGF
MM0326-5D20
Western Blot
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Bovine, Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
HGF
Human HGF
0.1 mL
Primary
3082