Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGF Mouse anti-Human, Bovine, Alexa Fluor™ 750, Clone: MM0326-5D20, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212355AF750

Catalog No. NP2123A70


Add to cart

Description

Description

HGF Monoclonal antibody specifically detects HGF in Human, Bovine samples. It is validated for Western Blot.
Specifications

Specifications

HGF
MM0326-5D20
Western Blot
Conjugated, Purified
HGF
Human HGF
Protein G purified
Cancer, Cell Cycle and Replication
Primary
3082.0
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Bovine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit