Learn More
HGFR/c-MET Mouse anti-Human, DyLight 755, Clone: 556, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243641IR
Catalog No. NBP243641IR
Add to cart
Description
Description
HGFR/c-MET Monoclonal antibody specifically detects HGFR/c-MET in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay.
Specifications
Specifications
HGFR/c-MET
556
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
Purified
MET
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human c-Met. The exact sequence is proprietary.
Protein A purified
Cancer, Cancer Stem Cells, Cell Cycle and Replication, Cellular Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
4233
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
AUTS9, c-Met, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, HGFR, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human