Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ HGF Recombinant Human Protein Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.

HGF Recombinant Human Protein

$377.00

Specifications

Common Name HGF Recombinant Human Protein
For Use With (Application) Mammalian cell culture
Formulation Lyophilized
Gene Alias HGF
Name HGF Recombinant Human Protein
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PHG0254 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Gibco™
PHG0254
10μg Each for $377.00
Add to cart
 
Description

Description

Recombinant Human Hepatocyte Growth Factor (HGF) is a bioactive protein intended for use in cell culture applications. HGF plays a role in positive regulation of cell proliferation.

  • High purity—no interference from other proteins or contaminants
  • High biological activity—more results with less protein
  • Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media

    High purity recombinant bioactive protein
    Recombinant Human Hepatocyte Growth Factor is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

    High biological activity
    In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Hepatocyte Growth Factor (HGF) specific activity is determined by the dose dependent proliferation of monkey 4MBr-5 cells.

    Proven compatibility
    Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

    See our selection of recombinant Gibco™ growth factors.

    For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Specifications

    Specifications

    HGF Recombinant Human Protein
    Lyophilized
    HGF Recombinant Human Protein
    HGF Recombinant Human Protein
    HGF (hepatocyte growth factor)
    Cell culture
    Human
    Mammalian cell culture
    HGF
    1 Each
    Growth factors and receptors
    For research use only. Not for use in diagnostic procedures
    Lyophilized

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit