Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHAT Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Supplier: Bioss BS15474R
Description
Skinny hedgehog′ (SKI1) encodes an enzyme that acts within the secretory pathway to catalyze amino-terminal palmitoylation of ′hedgehog′ (see MIM 600725).Specifications
HHAT | |
Polyclonal | |
Unconjugated | |
HHAT | |
Hedgehog acyltransferase; HHAT; MART2; MART-2; melanoma antigen recognized by T cells 2; Melanoma antigen recognized by T-cells 2; Protein-cysteine N-palmitoyltransferase HHAT; SKI1; Skinny hedgehog protein 1; Skn | |
Rabbit | |
Protein A | |
RUO | |
55733 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5VTY9 | |
HHAT | |
KLH conjugated synthetic peptide derived from human HHAT. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction