Learn More
Aviva Systems Biology Corp HHEX Antibody - C-terminal region (ARP58009_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58009_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HHEX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HHEX. Alternate names: HEX, HMPH, HOX11L-PEN, PRH, PRHXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62212