Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHEX Mouse anti-Human, Mouse, Rat, Alexa Fluor 488, Clone: OTI3E6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270890AF488
Description
HHEX Monoclonal specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
HHEX | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HHEX | |
Recombinant fragment expressed in E.coli corresponding to amino acids 100-270 of human HHEX. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI3E6 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor | |
Mouse | |
Affinity Purified | |
Developmental Biology | |
3087.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title