Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hHR23A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152942

 View more versions of this product

Catalog No. NBP152942


Add to cart

Description

Description

Rad23 Polyclonal antibody specifically detects Rad23 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

hHR23A
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
RAD23A
Synthetic peptides corresponding to RAD23A(RAD23 homolog A (S. cerevisiae)) The peptide sequence was selected from the middle region of RAD23A. Peptide sequence GIPGSPEPEHGSVQESQVSEQPATEAAGENPLEFLRDQPQFQNMRQVIQQ.
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Guinea pig: 92%.
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P54725
hHR23A, HHR23ARAD23, yeast homolog, A, HR23A, MGC111083, RAD23 (S. cerevisiae) homolog A, RAD23 homolog A (S. cerevisiae), UV excision repair protein RAD23 homolog A
Rabbit
IgG
100ul
Apoptosis, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors
Primary
5886
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit