Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hHR23A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15292020UL

 View more versions of this product

Catalog No. NBP15292020


Add to cart

Description

Description

hHR23A Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

hHR23A
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
RAD23A
Synthetic peptides corresponding to RAD23A(RAD23 homolog A (S. cerevisiae)) The peptide sequence was selected from the N terminal of RAD23A. Peptide sequence VPSSGSSGREEDAASTLVTGSEYETMLTEIMSMGYERERVVAALRASYNN.
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P54725
hHR23A, HHR23ARAD23, yeast homolog, A, HR23A, MGC111083, RAD23 (S. cerevisiae) homolog A, RAD23 homolog A (S. cerevisiae), UV excision repair protein RAD23 homolog A
Rabbit
IgG
20ul
Apoptosis, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors
Primary
5886
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit