Learn More
Cole-Parmer Megger 230415 AC Hipot Tester, 0-4 kV Applied Test Voltage  DFS Item
Manufacturer:  Cole-Parmer 2002264
These AC and AC/DC High-Pot Testers are designed for safety and ease of operation, and readily adapt to development, maintenance or production testing. Three models are available: two AC and one AC/DC. The AC High-Pot Testers feature a front-panel, NEMA 5-15IR three-prong output receptacle. Booted, alligator-clip output and return probes are also supplied with all models.Test voltage is continuously adjustable from zero to full output, with a zero-start interlock. Leakage current trip level may be adjusted from 0.3 to 12 mA by a front-panel knob. Push buttons are provided to select one-second or continuous testing, to turn off the high voltage and to reset the instrument. All high-voltage connections are recessed to prevent accidental contact with high voltage.
Catalog No. 50-631-579