Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Hi-Vis Nitrile-Coated Impact Resistant Work Gloves with Fleece Lining
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Impact resistant work gloves deliver an extra level of protection to the back of the hand and fingers
- Hi-vis yellow shell constructed of a synthetic fiber blend improves awareness on the job site
- Sandy nitrile coating on the palm provides a non-slip surface in wet, dry and oily conditions
- Fleece liner increases warmth in cold conditions
- Ideal for applications that require multi-hazard protection, grip, dexterity, and thermal protection
Specifications
Coating Material | Nitrile |
Color | Hi-vis Orange |
Cut Resistance Rating | ANSI A4 |
Hand | Specific |
Material | HPPE |
Size | Medium |
Glove Cuff Color | Hi-Vis Yellow |
Glove Cuff Style | Knit |
Liner Material | Fleece |
Series | I2449T |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction