Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Hi-Vis Safety Vests with Pockets

Pyramex™ Hi-Vis Safety Vests with Pockets meet ANSI/ISEA 107-2015 safety standards

Manufacturer:  Pyramex™ RVZ2110X2

 View more versions of this product

Catalog No. 19-555-494

This item is not returnable. View return policy

Description

Description

  • Zipper closure
  • One 2 in. silver reflective stripe applied horizontally across lower chest
  • Two 2 in. silver reflective stripes run vertically from top of horizontal stripes, over shoulders to top of back side horizontal stripe
  • Outside left chest pocket measures 5.5 x 6.5 in.
  • Inside right patch pocket measures 7.5 x 7.5 in.
Specifications

Specifications

ANSI/ISEA 107-2015, Type R - Class 2
Hi-Vis Lime
Polyester Mesh
2
Zipper
2X-Large
50 to 52 in.
China Brand Silver
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit