Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ Hi-Vis Crinkle Latex Coated Seamless Knit Acrylic Gloves

Textured latex coating enhances grip capabilities in wet or dry conditions, in addition to providing abrasion resistance. Acylic material provides economical cold-weather protection for refrigeration, food processing, and outdoor winter activities.

Specifications

Color Hi-Vis Yellow, Black
Coating Material Latex
For Use With (Application) Refrigeration, Food Processing, Commercial Fishing, and Construction
Hand Specific
Material Nylon
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-046-402
View Documents
PIP™
411420/L
Large
Dozen of 12 for $75.30
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-046-403
View Documents
PIP™
411420/XL
X-Large
Dozen of 12 for $75.30
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-046-400
View Documents
PIP™
411420/S
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
19-046-401
View Documents
PIP™
411420/M
Medium This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description

Description

  • Acrylic material provides economical cold weather protection
  • Latex coating enhances grip capabilities in wet/dry conditions
  • Knit wrist prevents dirt and debris from entering the glove
  • Hi-vis coloring increases worker safety in low-light environments
  • Gloves quickly evaporate moisture and perspiration from the skin
  • Launderable fabric extends glove life and reduce replacement costs
Specifications

Specifications

Hi-Vis Yellow, Black
Refrigeration, Food Processing, Commercial Fishing, and Construction
Nylon
41-1420
Latex
Specific
Knit
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit