Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ M-Pact™ E5 Impact Resistant Gloves with D30™ Padding

Catalog Number 19030114
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

These highly visible gloves feature thermoplastic rubber to absorb heavy impacts, and D30™ palm padding to dissipate high-impact energy.

  • Thermoplastic Rubber (TPR) pinch-point fingertip protection
  • High-visibility TrekDry™ keeps hands cool and comfortable
  • Reflective print provides eye-catching luminance in low-light environments
  • D3O™ palm padding absorbs and dissipates high-impact energy
  • Armortex™ palm reinforcement provides added durability
  • Reinforced index finger and thumb deliver durability where its needed most
  • Hook and Loop closure provides a secure fit
  • Touchscreen capable synthetic leather to stay connected while on the job
  • Machine washable
ANSI Abrasion Rating ANSI 4
Back Material TrekDry™
Color Hi-vis Yellow
Cut Resistance Rating ANSI A5
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size 2X-Large
Finger Style Full
For Use With (Application) Construction, Heavy Equipment Operation, Towing, and Transportation
Glove Cuff Style Hook and Loop
Liner Material Armortex™
Series M-Pact™ E5
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit