Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIBADH Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15472220UL

 View more versions of this product

Catalog No. NBP15472220


Add to cart

Description

Description

HIBADH Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

HIBADH
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
HIBADH
Synthetic peptides corresponding to HIBADH(3-hydroxyisobutyrate dehydrogenase) The peptide sequence was selected from the middle region of HIBADH. Peptide sequence AKEVEKMGAVFMDAPVSGGVGAARSGNLTFMVGGVEDEFAAAQELLGCMG.
35 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
P31937
3-hydroxyisobutyrate dehydrogenase, 3-hydroxyisobutyrate dehydrogenase, mitochondrial, EC 1.1.1,3'-hydroxyisobutyrate dehydrogenase, EC 1.1.1.31, MGC40361, NS5ATP1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
11112
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit