Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 100-4.6
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 100-4.6 come with customized packing (please specify packing material) and are 100mmL x 4.6mmI.D.
$673.65 - $1291.24
Specifications
Column Type Hibar™ HPLC  column
Length (Metric) 100mm
Diameter (Metric) Inner 4.6mm
Sorbent Customized (please specify packing material)
View More Specs
Products 4
Catalog Number Mfr. No. Stationary Phase Mobile Phase Compatibility Price Quantity & Availability  
Catalog Number Mfr. No. Stationary Phase Mobile Phase Compatibility Price Quantity & Availability  
M1500137004 View Documents MilliporeSigma™
1.50013.7004
HPLC RP (reversed-phase) Each for $1,291.24
Add to cart
M1500137052 View Documents MilliporeSigma™
1.50013.7052
RP18 RP (reversed-phase) Each for $708.69
Add to cart
M1500137076 View Documents MilliporeSigma™
1.50013.7076
NH2 RP (reversed-phase, when in mode) & NP (normal-phase) Each for $708.69
Add to cart
M1500137109 View Documents MilliporeSigma™
1.50013.7109
60SI NP (normal-phase) Each for $673.65
Add to cart
Specifications
Specifications
Hibar™ HPLC  column
100mm
4.6mm
Customized (please specify packing material)