Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 125-4
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma™ Hibar™ HPLC Columns, 125-4 come with customized packing (please specify packing material) and are 125mmL x 4mmI.D.
Manufacturer:  MilliporeSigma™ 1.50181.7087
Catalog No. M1501817087
Add to cart
Description
Description
Specifications
Specifications
4 mm
RP (reversed-phase)
Customized (please specify packing material)
125 mm
RP8
Hibar™ HPLC Column