Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 125-4
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 125-4 come with customized packing (please specify packing material) and are 125mmL x 4mmI.D.
$673.65 - $1070.26
Specifications
Column Type Hibar™ HPLC  column
Length (Metric) 125mm
Diameter (Metric) Inner 4mm
Sorbent Customized (please specify packing material)
View More Specs
Products 9
Catalog Number Mfr. No. Stationary Phase Mobile Phase Compatibility Price Quantity & Availability  
Catalog Number Mfr. No. Stationary Phase Mobile Phase Compatibility Price Quantity & Availability  
M1501817071 View Documents MilliporeSigma™
1.50181.7071
CN5 RP (reversed-phase, when in mode) & NP (normal-phase) Each for $708.69
Add to cart
M1501817076 View Documents MilliporeSigma™
1.50181.7076
NH2 RP (reversed-phase, when in mode) & NP (normal-phase) Each for $708.69
Add to cart
M1501817078 View Documents MilliporeSigma™
1.50181.7078
HPLC RP (reversed-phase) Each for $1,070.26
Add to cart
M1501817080 View Documents MilliporeSigma™
1.50181.7080
Each for $684.00
Add to cart
M1501817109 View Documents MilliporeSigma™
1.50181.7109
60SI NP (normal-phase) Each for $673.65
Add to cart
M1501817116 View Documents MilliporeSigma™
1.50181.7116
RP18 RP (reversed-phase) Each for $940.88
Add to cart
M1501817140 View Documents MilliporeSigma™
1.50181.7140
RP8 RP (reversed-phase) Each for $884.83
Add to cart
M1501817141 View Documents MilliporeSigma™
1.50181.7141
RPSELECTB RP (reversed-phase) Each for $973.95
Add to cart
M1501817174 View Documents MilliporeSigma™
1.50181.7174
SI3 NP (normal-phase) Each for $955.93
Add to cart
Specifications
Specifications
Hibar™ HPLC  column
125mm
4mm
Customized (please specify packing material)