Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIBCH Mouse anti-Human, Canine, Allophycocyanin/Cy7, Clone: OTI3H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271796APCCY7

Catalog No. N271796APC7


Add to cart

Description

Description

HIBCH Monoclonal antibody specifically detects HIBCH in Human, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

HIBCH
OTI3H5
Flow Cytometry
Affinity Purified, Conjugated
HIBCH
Full length recombinant protein of human HIBCH (NP_055177) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
26275.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
3-hydroxyisobutyryl-CoA hydrolase, EC 3.1.2.4,3-hydroxyisobutyryl-coenzyme A hydrolase3-hydroxyisobutyryl-CoA hydrolase, mitochondrial, HIB-CoA hydrolase, HIBYL-CoA-H, HIBYLCOAH
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit