Learn More
HIBCH Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI4G1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525155
Catalog No. PIMA525155
Add to cart
Description
Description
Beta-hydroxyisobutyryl-CoA hydrolase (EC 3.1.2.4) is responsible for the specific hydrolysis of HIBYL-CoA, a valine catabolite, as well as the hydrolysis of beta-hydroxypropionyl-CoA, an intermediate in a minor pathway of propionate metabolism.
Specifications
Specifications
HIBCH
Monoclonal
0.7 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6NVY1, Q5XIE6
HIBYLCOAH, 3-hydroxyisobutyryl-CoA hydrolase, mitochondrial, 3-hydroxyisobutyryl-Coenzyme A hydrolase, HIB-CoA hydrolase, HIBYL-CoA-H
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26275, 301384, 607040, 713951
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI4G1
Unconjugated
HIBCH
Liquid
Hibch
Full length human recombit protein of HIBCH produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat