Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HICE1 Mouse anti-Human, Clone: 15A2-35, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527860

Catalog No. PIMA527860

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Positive Control: mouse adipose Predicted Reactivity: Human (100%), Rat (92%), Pig (92%), Sheep (94%), Bovine (94%)

Hice1 contributes to mitotic spindle assembly, maintece of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
Specifications

Specifications

HICE1
Monoclonal
1 mg/ml
PBS with no preservative
Q9BT25
MGC20533, NYSAR48, HICE1, HAUS8, NY-SAR-48, NY SAR 48
Mouse
IgG1
100 μl
-20° C, Avoid Freeze/Thaw Cycles
Primary
93323
Western Blot
15A2-35
Unconjugated
HAUS8
Liquid
HAUS8
Human Hice1
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit