Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HID1 Mouse anti-Human, Rat, Clone: OTI2E9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525164

Catalog No. PIMA525164


Add to cart

Description

Description

CQ028 may play an important role in the development of cancers in a broad range of tissues.
Specifications

Specifications

HID1
Monoclonal
0.59 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IV36
DMC1, UPF0663 transmembrane protein C17orf28, down-regulated in multiple cancers 1, downregulated in multiple cancer 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283987, 287822
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2E9
Unconjugated
HID1
Liquid
Hid1
Full length human recombit protein of C17orf28 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit