Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Mouse anti-Human, Mouse, PE, Clone: Mgc3, eBioscience Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 12752880

 View more versions of this product

Catalog No. 12-752-880

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Description: This Mgc3 monoclonal antibody detects hypoxia-inducible factor 1 alpha (HIF-1 alpha) from human, non-human primate, bovine, mouse and porcine cells. This antibody does not cross-react with ARNT or the related HIF-2 alpha. Mgc3 has been successfully used in western blotting, immunofluorescence, immunocytochemistry, immunoprecipitation, flow cytometry and gel shift procedures. By western blot, this antibody detects a ~116 kDa protein representing HIF-1 alpha after hypoxic induction in COS cells. Immunofluorescence staining of HIF-1 alpha in COS-7 cells with clone Mgc3 yields nuclear staining after exposing cells to 1% oxygen for 4 hours. In gel shift assay experiments, the Mgc3 antibody detected only mouse and not human HIF-1 alpha. Applications Reported: This Mgc3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This Mgc3 antibody has been tested by flow cytometric analysis of DFOA stimulated HeLa cells using the methanol fixation protocol. This may be used at less than or equal to 1.0 µg/mL per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

HIF1 alpha functions as a transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, HIF-1 activates the transcription of over 40 genes, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, cell proliferation, tumor angiogenesis, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. HIF-1 alpha is regulated post-translationally by Factors Inhibiting HIF (FIH) and Prolyl Hydroxylase (PHD’s). Once accumulated HIF1 alpha translocates into the nucleus and binds to its heterodynamic binding partners, ARNT and p300, facilitating expression of genes involved in the hypoxic response. Dysregulation of HIF1 alpha signaling has been observed in several tumor types. Hypoxia contributes significantly to the pathophysiology of multiple human diseases, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease, and chronic obstructive pulmonary disease.
Specifications

Specifications

HIF-1 alpha
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q16665, Q61221
HIF1; MOP1; PASD8; HIF-1A; bHLHe78; HIF-1alpha; HIF1-ALPHA; HIF-1-alpha
Mouse
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
15251, 3091
Flow Cytometry
Mgc3
PE
HIF1A
Liquid
HIF1A
Human HIF-1 alpha amino acids 530-826.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit