Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Rabbit anti-Human, DyLight™ 350, Clone: 2443B, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275977UV

Catalog No. NBP275977UV


Add to cart

Description

Description

HIF-1 alpha Monoclonal antibody specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay.
Specifications

Specifications

HIF-1 alpha
2443B
Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated
Conjugated, Purified
HIF1A
This recombinant HIF-1 alpha Antibody (2443B) was developed against partial recombinant human HIF-1 alpha protein (amino acids 1-826) [Uniprot# Q16665].
Protein A or G purified
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary
3091.0
Immunoassay, Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit