Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF1AN Mouse anti-Human, Mouse, Clone: FIH 162c, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527619

Catalog No. PIMA527619


Add to cart

Description

Description

Hydroxylates HIF-1 alpha at  ft.Asp-803 ft. in the C-terminal transactivation domain (CAD). Functions as an oxygen sensor and, under normoxic conditions, the hydroxylation prevents interaction of HIF-1 with transcriptional coactivators including Cbp/p300-interacting transactivator. Involved in transcriptional repression through interaction with HIF1A, VHL and histone deacetylases. Hydroxylates specific Asn residues within ankyrin repeat domains (ARD) of NFKB1, NFKBIA, NOTCH1, ASB4, PPP1R12A and several other ARD-containing proteins. Also hydroxylates Asp and His residues within ARDs of ANK1 and TNKS2, respectively. Negatively regulates NOTCH1 activity, accelerating myogenic differentiation. Positively regulates ASB4 activity, promoting vascular differentiation.
Specifications

Specifications

HIF1AN
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Q8BLR9, Q9NWT6
Factor inhibiting HIF1 (hypoxia-inducible factor), DKFZp762F1811, Factor inhibiting HIF-1, Factor inhibiting HIF1, FIH 1, FIH-1, FIH1, FLJ20615, FLJ22027, HIF1AN, HIF1N_HUMAN, Hypoxia inducible factor 1 alpha inhibitor, Hypoxia inducible factor 1 alpha subunit inhibitor, Hypoxia inducible factor asparagine hydroxylase, Hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, Peptide aspartate beta dioxygenase
Mouse
IgG1
100 μg
-20°C
Primary
319594, 55662
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
FIH 162c
Unconjugated
HIF1AN
Liquid
HIF1AN
Full length human FIH expressed in E.coli BL21 (DE3) cells
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit