Learn More
Fisherbrand™ High Bench Height Blue Polyurethane Chair with Medium Back Products for Science Education
Fisherbrand High Bench Height Blue Polyurethane Chair with Medium Back features industrial grade, puncture, soil, and chemical resistant polyurethane seat and back. Fully adjustable for high 42 in high counters.
$267.00 - $367.00
Specifications
Adjustable (Height) 23 to 33 in
Base Style BLACK
Capacity 300 lbs
Color BLUE
Seating Form HIGH
View More Specs
Products 8
Catalog Number Arm Style With Armrest With Tilt With Casters Price Quantity & Availability  
Catalog Number Arm Style With Armrest With Tilt With Casters Price Quantity & Availability  
S40401 Products for Science Education View Documents Adjustable YES NO NO Each for $326.00
Add to cart
S40404 Products for Science Education View Documents Adjustable YES YES NO Each for $355.00
Add to cart
S40411 Products for Science Education View Documents Adjustable YES NO YES Each for $339.00
Add to cart
S40414 Products for Science Education View Documents Adjustable YES YES YES Each for $367.00
Add to cart
S40418 Products for Science Education View Documents Not adjustable NO NO YES Each for $280.00
Add to cart
S40419 Products for Science Education View Documents Not adjustable NO NO NO Each for $267.00
Add to cart
S40422 Products for Science Education View Documents Not adjustable NO YES YES Each for $306.00
Add to cart
S40426 Products for Science Education View Documents Not adjustable NO YES NO Each for $296.00
Add to cart
Description
Description
  • Features ergonomic adjustments
  • Larger Back Pad for extra comfort
  • Optional 6-way adjustable arms for proper ergonomic arm placement
  • Optional 8-degree forward, 3-degreee backward locking seat tilt
  • For 42 in high work surface
  • 20-Inch diameter chrome foot ring
  • Blue, Soil, Puncture, and Chemical Resistant Industrial Grade Polyurethane Seat and Back
  • Non-Marring Casters or Stationary Glides
  • Meets/exceeds ANSI/BIFMA standards
  • 15-year warranty, lifetime warranty on pneumatic lift
Certifications
ANSI/BIFMA
Warranty and Services
15-year on parts, lifetime on pneumatic cylinder
Specifications
Specifications
23 to 33 in
300 lbs
HIGH
PNEUMATIC
YES
BLACK
BLUE
HIGH BENCH
POLYURETHANE
NO