Learn More
Fisherbrand™ High Bench Height Class 100 Clean Room Chair with Chrome Foot Ring and Adjustable Arms Products for Science Education
Fisherbrand High Bench Height Class 100 Clean Room Chair with Chrome Foot Ring and Adjustable Arms features antimicrobial/antibacterial vinyl. Fully adjustable for high 42 in counters.
$428.00 - $456.00
Specifications
Adjustable (Height) 26 to 35.5 in
Base Style CHROME
Capacity 300 lbs
Lift Type PNEUMATIC
Material VINYL
View More Specs
Products 20
Catalog Number With Tilt Color With Casters With Armrest Price Quantity & Availability  
Catalog Number With Tilt Color With Casters With Armrest Price Quantity & Availability  
S40164 Products for Science Education View Documents NO Blue YES YES Each for $428.00
Add to cart
S40165 Products for Science Education View Documents NO Blue NO YES Each for $428.00
Add to cart
S40166 Products for Science Education View Documents YES Blue YES YES Each for $456.00
Add to cart
S40172 Products for Science Education View Documents YES Blue NO YES Each for $456.00
Add to cart
S40179 Products for Science Education View Documents NO BLACK YES YES Each for $428.00
Add to cart
S40188 Products for Science Education View Documents NO BLACK NO YES Each for $428.00
Add to cart
S40190 Products for Science Education View Documents YES BLACK YES YES Each for $456.00
Add to cart
S40191 Products for Science Education View Documents YES BLACK NO YES Each for $456.00
Add to cart
S40198 Products for Science Education View Documents NO BURGUNDY YES YES Each for $428.00
Add to cart
S40205 Products for Science Education View Documents YES BURGUNDY NO YES Each for $456.00
Add to cart
S40221 Products for Science Education View Documents YES Gray YES YES Each for $456.00
Add to cart
S40223 Products for Science Education View Documents YES Gray NO YES Each for $456.00
Add to cart
S40228 Products for Science Education View Documents NO Navy YES YES Each for $428.00
Add to cart
S40230 Products for Science Education View Documents YES Navy YES YES Each for $456.00
Add to cart
S40199 Products for Science Education View Documents NO BURGUNDY NO YES Each for $428.00
Add to cart
S40201 Products for Science Education View Documents YES BURGUNDY YES YES Each for $456.00
Add to cart
S40219 Products for Science Education View Documents NO Gray YES YES Each for $428.00
Add to cart
S40220 Products for Science Education View Documents NO Gray NO YES Each for $428.00
Add to cart
S40229 Products for Science Education View Documents NO Navy NO YES Each for $428.00
Add to cart
S40231 Products for Science Education View Documents YES Navy NO YES Each for $456.00
Add to cart
Description
Description
  • Features ergonomic adjustments 
  • Larger Back Pad for extra comfort
  • Optional 8-degree forward, 3-degreee backward locking seat tilt
  • Optional 6-way adjustable arms for proper ergonomic arm placement
  • For 42 in high work surface
  • 20-Inch diameter chrome foot ring
  • Suitable for Class 100 (ISO Class 5) Clean Rooms
  • Antimicrobial/antibacterial vinyl for easy cleaning
  • Self-braking, dual-wheel casters or non-marring stationary glides suitable for hard floors
Certifications
ANSI/BIFMA
Warranty and Services
15-year on parts, 5-year on upholstery, Lifetime on the pneumatic lift
Specifications
Specifications
26 to 35.5 in
300 lbs
VINYL
YES
HIGH BENCH
CHROME
PNEUMATIC
YES
HIGH
Adjustable