Learn More
Fisherbrand™ High Bench Height Class 100 Industrial Chrome and Polyurethane Clean Room Chair Chrome with Adjustable Arms Products for Science Education
Fisherbrand High Bench Height Class 100 Industrial Chrome and Polyurethane Clean Room Chair Chrome with Adjustable Arms features industrial grade, puncture, soil, and chemical resistant seat and back. Fully adjustable for high 42 in high counters.
$393.00 - $447.00
Specifications
Seating Height HIGH BENCH
Base Style CHROME
Capacity 300 lbs
Lift Type PNEUMATIC
Material POLYURETHANE
View More Specs
Products 8
Catalog Number Color With Casters With Tilt Price Quantity & Availability  
Catalog Number Color With Casters With Tilt Price Quantity & Availability  
S40352 Products for Science Education View Documents BLACK NO NO Each for $393.00
Add to cart
S40353 Products for Science Education View Documents BLACK YES NO Each for $393.00
Add to cart
S40354 Products for Science Education View Documents BLACK YES YES Each for $421.00
Add to cart
S40355 Products for Science Education View Documents BLACK NO YES Each for $421.00
Add to cart
S41413 Products for Science Education View Documents BLUE NO NO Each for $418.00
Add to cart
S41414 Products for Science Education View Documents BLUE YES NO Each for $418.00
Add to cart
S41415 Products for Science Education View Documents BLUE YES YES Each for $447.00
Add to cart
S41416 Products for Science Education View Documents BLUE NO YES Each for $447.00
Add to cart
Description
Description
  • Features ergonomic adjustments
  • 6-way adjustable arms for proper ergonomic arm placement
  • Optional 8-degree forward, 3-degreee backward locking seat tilt
  • Handle on back pad for ease of movement around the work space
  • For 42 in high work surface
  • Suitable for Class 100 (ISO Class 5) Clean Rooms
  • 20 in Diameter Chrome Adjustable Foot Ring
  • Soil, Puncture, and Chemical Resistant Industrial Grade Polyurethane Seat and Back
  • Non-Marring Casters or Stationary Glides
Certifications
ANSI/BIFMA
Warranty and Services
15-year on parts, lifetime on pneumatic cylinder
Specifications
Specifications
HIGH BENCH
300 lbs
POLYURETHANE
NO
YES
CHROME
PNEUMATIC
YES
Adjustable