Learn More
Fisherbrand™ High Bench Height Industrial Blue Polyurethane Chrome Chair Products for Science Education
Fisherbrand High Bench Height Industrial Blue Polyurethane Chrome Chair features industrial grade, puncture, soil, and chemical resistant seat and back. Fully adjustable for high 42 in high counters.
$297.00 - $404.00
Specifications
Seating Height HIGH BENCH
Capacity 300 lbs
Lift Type PNEUMATIC
Material POLYURETHANE
With Backrest YES
View More Specs
Products 8
Catalog Number With Tilt With Casters With Armrest Arm Style Price Quantity & Availability  
Catalog Number With Tilt With Casters With Armrest Arm Style Price Quantity & Availability  
S40491 Products for Science Education View Documents NO NO YES ADJUSTABLE Each for $377.00
Add to cart
S40492 Products for Science Education View Documents NO YES YES ADJUSTABLE Each for $377.00
Add to cart
S40496 Products for Science Education View Documents YES YES YES ADJUSTABLE Each for $404.00
Add to cart
S40500 Products for Science Education View Documents YES NO YES ADJUSTABLE Each for $404.00
Add to cart
S40510 Products for Science Education View Documents NO NO NO Non adjustable Each for $297.00
Add to cart
S40511 Products for Science Education View Documents NO YES NO Non adjustable Each for $297.00
Add to cart
S40519 Products for Science Education View Documents YES NO NO Non adjustable Each for $325.00
Add to cart
S40520 Products for Science Education View Documents YES YES NO Non adjustable Each for $325.00
Add to cart
Description
Description
  • Features ergonomic adjustments
  • Optional 6-way adjustable arms for proper ergonomic arm placement
  • Optional 8-degree forward, 3-degreee backward locking seat tilt
  • Handle on back pad for ease of movement around the work space
  • For 42 in high work surface
  • 20 in Diameter Chrome Adjustable Foot Ring
  • Soil, Puncture, and Chemical Resistant Industrial Grade Polyurethane Seat and Back
  • Non-Marring Casters or Stationary Glides
Certifications
ANSI/BIFMA
Warranty and Services
15-year on parts, lifetime on pneumatic cylinder
Specifications
Specifications
HIGH BENCH
PNEUMATIC
YES
HIGH
300 lbs
POLYURETHANE
NO
CHROME