Learn More
Brady™ High Density Polypropylene Danger Sign: HAZARDOUS MATERIAL
Improve workplace safety with proper signage. The Brady™ Danger Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$28.90
Specifications
Shape | Rectangular |
---|---|
Displayed Text | HAZARDOUS MATERIAL |
Temperature Range (English) | -40°F to 185°F |
Finish | Not Laminated |
Material | Polypropylene |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-104-884
|
Brady™
116154 |
14 x 10 in. |
Each for $28.90
|
|
|||||
19-104-885
|
Brady™
116155 |
10 x 7 in. |
Each for $28.90
|
|
|||||
Description
- High density polypropylene material is abrasion-resistant and chemical-resistant
- Bold and bright, providing high visibility to critical messages
Specifications
Rectangular | |
-40°F to 185°F | |
Polypropylene | |
Non-adhesive | |
Corner holes | |
-40°F |
HAZARDOUS MATERIAL | |
Not Laminated | |
1 Ea. | |
High density polypropylene | |
185°F | |
0.0551 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.