Learn More
Brady™ High Density Polypropylene Sign: CAUTION - HOT
Improve workplace safety with proper signage and markings. This Brady™ high density polypropylene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 116232
 View more versions of this product
Catalog No. 19-104-894
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • High density polypropylene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Safety
PP (Polypropylene)
14 in. x 10 in. x 0.0551 in.
Black on Yellow
Rectangle
CAUTION - HOT