Learn More
Research Diets IncSupplier Diversity Partner HIGH FAT DIET 60%KCAL FAT-100K DFS Item
Manufacturer:  Research Diets IncSupplier Diversity Partner D12492I100KG
Rodent Diet with 60% kcal% fat. Used in Research for Obesity and Diabetes. Product is packed in 12.5 kg box. Gamma-Irradiation. Pellet, Powder, Liquid Shelf Life. Most diets require storage in a cool dry environment. Stored correctly they should last 3-6 months. Because of the high fat content is best if kept frozen.
Catalog No. NC0064127