Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ A+ Aloe™ N88 Nitrile Gloves

Robust aloe and chamomile coated nitrile gloves offer comfortable protection for a variety of industries including healthcare, life science, manufacturing, machinery and equipment.

$19.05

Specifications

Applications Healthcare, Laboratory, Industrial
AQL 1.5
Certifications/Compliance ASTM D6319, FDA21 CFR 177-2600
Color Green
Thickness (English) 4.7 mil
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-170-805
View Documents Promotion Details
Ansell
N88110
Small
Pack of 100 for $19.05
Case of 10 PK for $190.48
Add to cart
 
19-170-806
View Documents Promotion Details
Ansell
N88210
Medium
Pack of 100 for $19.05
Case of 10 PK for $190.48
Edge
Add to cart
 
19-170-807
View Documents Promotion Details
Ansell
N88310
Large
Pack of 100 for $19.05
Case of 10 PK for $190.48
Add to cart
 
19-170-808
View Documents Promotion Details
Ansell
N88410
X-Large
Pack of 100 for $19.05
Case of 10 PK for $190.48
Edge
Add to cart
 
19-170-809
View Documents Promotion Details
Ansell
N88510
2X-Large
Pack of 100 for $19.05
Case of 10 PK for $190.48
Add to cart
 
Description

Description

  • Inner coating of aloe helps protect hands from chapping, drying and cracking
  • Ideal for extended wear
  • Textured fingertip finish provides a secure grip
  • 100% nitrile construction offers abrasion, chemical and puncture resistance
Specifications

Specifications

Healthcare, Laboratory, Industrial
ASTM D6319, FDA21 CFR 177-2600
4.7 mil
Partially Textured
5.9 mil
4.7 mil
1.5
Green
0.12 mm
16 MPa
0.15 mm
0.12 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit