Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microflex™ ALOE SENSE™ Nitrile Exam Gloves

Aloe coated, powder free nitrile exam glove provides extra sensitivity with soothing aloe.

Manufacturer:  Microflex™ N964

 View more versions of this product

Catalog No. 19-170-828



Add to cart

Description

Description

  • Extra soft nitrile exam glove
  • Offers tactile sensitivity and comfort
  • Soothing aloe helps protect the hands from drying and cracking
  • Fully textured for an excellent grip
Specifications

Specifications

Textured finger
Beaded/Rolled
Medical, exam
Nitrile
20
Powder-free
Non-sterile
24.13 cm
Yes
Ambidextrous
Yes
X-Large
3 mil
Blue, Green
9.5 in.
AQL 1.5, ISO9001 QMS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit