Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell Cobalt™ Ultra Nitrile Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
Manufactured using an ultra-soft and stretchy nitrile formulation for excellent durability and comfort. Its thin construction results in enhanced tactile sensitivity, while textured fingers provide a firm grip. Smartly packed 200 gloves per box to save money and reduce environmental waste.
Specifications
Certifications/Compliance | AQL 1.5, FDA, ASTM F 1671, ISO9001 |
Color | Blue |
Disposable | Single-use |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | Medium |
Thickness (English) | 3 mil |
Thickness (Metric) | 0.07 mm |
Tensile Strength | 20.5 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction