Learn More
High Five™ Facemasks
Ideal for various medical or lab procedures
Manufacturer:  High Five™ AM101H
 View more versions of this product
Catalog No. 19-167-899
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Non-woven
  • 3-ply masks are designed for comfort
Specifications
Specifications
Blue
Yes
Yes
3-ply
PP (Polypropylene)
95% (BFE)
Earloops, Heavy duty
Sonic welded Spunbond Polypropylene; Non-woven