High Five™ Industrial Latex Gloves

Offer exceptional dexterity

Manufacturer: Microflex Corporation L564

 View more versions of this product

Catalog No. 19-167-07

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

This product contains natural rubber latex, a component of which may cause an allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of this product.


    Specifications

    Glove Cuff Style Beaded
    For Use With (Application) food processing and other industrial areas
    Length (English) 9.5 in.
    Material Latex
    Coating Material Textured
    Length (Metric) 24cm
    Powdered No
    Size X-Large
    Sterile No
    Thickness 4.5mil
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit