Learn More
High Five™ Microporous Coveralls
Protection from light splash and particulates
Manufacturer:  High Five™ EW20103M
 View more versions of this product
Catalog No. 19-167-901
Add to cart
Description
Description
Ideal for general-purpose applications such as maintenance and general cleanup, including oily applications.
  • Microporous construction for breathablity and comfort
  • Full cut for improved fit and comfort
  • Zipper closure
  • Elastic wrists and ankles
  • White
Specifications
Specifications
Elastic
Medium
Elastic wrists and ankles
White
Microporous Coverall