Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
High Five™ Microporous Lab Coats
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Description
Breathability with durability
- Cloth-like, microporous material
- General purpose protection from dirt, grime, splashes and spills
- Economical and lightweight
- Low linting and antistatic
- Elastic wrists, five-snap closure and two pockets
- White
Specifications
Size | 2X-Large |
Material | Microporous Film |
Color | White |
Closure Type | Snaps |
No. of Pockets | 2 |
For Use With (Application) | General purpose protection from dirt, grime, splashes and spills |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction