High Five Nitrile Disposable Gloves

Economical alternative to latex gloves for industrial and food-grade applications

Manufacturer: Microflex Corporation N204

 View more versions of this product

Catalog No. 19-167-053

  Check Availability
Add to Cart

    Specifications

    Certifications FDA, ISO9002
    Glove Cuff Style Beaded
    Hand Ambidextrous
    Grade Industrial, food
    Length (English) 9.5 in.
    Length (Metric) 24cm
    Material Nitrile
    Powdered No
    Size X-Large
    Sterile No
    Thickness 4mil
    Coating Material Textured
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit