High Five Powder-Free Latex Gloves with Aloe

Manufacturer: Microflex Corporation L933

 View more versions of this product

Catalog No. 19-170-817

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Certifications ISO 9001
    Glove Cuff Style Beaded
    Hand Ambidextrous
    Grade Medical/exam
    Length (English) 9.5 in.
    Length (Metric) 50cm
    Material Nitrile
    Powdered No
    Size Large
    Sterile No
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit